What Does DNA Look Like?
Leverandør: EDVOTEK
S-10
EDVOS-10EA 775 NOK
EDVOS-10
What Does DNA Look Like?
Lærings materiell Læremateriale for biologi Genetics Learning Activities
This lab activity shows students what real chromosomal DNA looks like and allows them to explore the procedures involved in DNA extraction.
  • For 10 lab groups
  • Complete in 30 minutes
Also required: 95% ethanol or isopropyl alcohol.
Leveringsinformasjon: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, minilinks, glass rod, DNA spooling rods, test tubes, and salt.