Experimentation kits, how clean is the water we drink and the air we breathe?
Leverandør: EDVOTEK
S-30
EDVOS-30EA 970 NOK
EDVOS-30
Experimentation kits, how clean is the water we drink and the air we breathe?
Lærings materiell Læremateriale for geofag Natural Resources Learning Activities
With this kit, students sample water and air and then grow any microbes present overnight. A safe and simple way to teach pollution.
  • For 10 lab groups
  • Complete in 30 minutes and grow overnight
Also required: Microscope with 400× magnification.
Leveringsinformasjon: Kit includes instructions, multispot slides (4 cell types each), eosin and methylene blue stain, mounting medium, cover slips, transfer pipettes, and immersion troughs.