Experimentation kits, how does a doctor test for AIDS?
Leverandør: EDVOTEK
S-70
EDVOS-70EA 1075 NOK
EDVOS-70
Experimentation kits, how does a doctor test for AIDS?
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
The body defends itself from attack by infectious agents like bacteria and viruses by producing antibodies. Enzyme linked immunosorbent assays (ELISA) test for antibodies present in the blood, which indicate infection. In this experiment, students perform a simulated ELISA test to identify infected samples and compare them to control samples.
  • For 10 lab groups
  • Complete in 45 minutes
Leveringsinformasjon: Kit includes instructions, antigens, positive and negative controls, sera, secondary antibody, substrate, detection strips, transfer pipettes, and test tubes.