Experimentation kits, whose fingerprints were left behind?
Leverandør: EDVOTEK
S-91
EDVOS-91EA 1125 NOK
EDVOS-91
Experimentation kits, whose fingerprints were left behind?
Lærings materiell Læremateriale for kriminologi Rettsmedisinsk sett
Evidence left behind at a crime scene can identify a potential culprit. Even in this age of DNA, fingerprints and blood stains are still important in helping to identify a criminal. In this experiment, students solve a crime by dusting for fingerprints and use fluorescent dust to search for and identify trace amounts of blood.
  • For 10 lab groups
  • Complete in 50 minutes
Also required: Alcohol and longwave UV light.
Leveringsinformasjon: Kit includes instructions, brushes, magnifying lens, fingerprint cards, black dusting powder, fluorescent green andMore Product Information