Amphotericin B

Leverandør: EHRENSTORFER
Warning

Synonymer: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

C10243050
EHERC10243050EA 4500 NOK
EHERC10243050
Amphotericin B
Amphotericin B
Formel: C₄₇H₇₃NO₁₇
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00877763
CAS-nummer: 1397-89-3
EINECS: 215-742-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR