1-klor-4-nitrobenzen

Leverandør: EHRENSTORFER
Danger

Synonymer: 4-Chloronitrobenzene

C11453700
EHERC11453700EA 1850 NOK
EHERC11453700
1-klor-4-nitrobenzen
1-klor-4-nitrobenzen
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
Kokepunkt: 239 °C (1013 hPa)
Smeltepunkt: 83 °C
Tetthet: 1,52 g/cm³ (20 °C)
Flash punkt: 127 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007285
CAS-nummer: 100-00-5
EINECS: 202-809-6
FN: 1578
ADR: 6.1,II
Merck Indeks: 13,02170

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR