2-Klor-4-nitrotoluen

Leverandør: EHRENSTORFER

C11456100
EHERC11456100EA 1475 NOK
EHERC11456100
2-Klor-4-nitrotoluen
2-Klor-4-nitrotoluen
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokepunkt: 260 °C (1013 hPa)
Smeltepunkt: 68 °C
MDL nummer: MFCD00007210
CAS-nummer: 121-86-8
EINECS: 204-501-7
FN: 3457
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR