2,2-Dichloropropionic acid

Leverandør: EHRENSTORFER

C11949500
EHERC11949500EA 2700 NOK
EHERC11949500
2,2-Dichloropropionic acid
2,2-Dichloropropionic acid
Formel: C₃H₄Cl₂O₂
MW: 142.97 g/mol
MDL nummer: MFCD00004178
CAS-nummer: 75-99-0
EINECS: 200-923-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR