Difenylamin

Leverandør: EHRENSTORFER
Danger

Synonymer: N-Phenylaniline, N-Phenylbenzeneamine

C12890000
EHERC12890000EA 885 NOK
EHERC12890000
Difenylamin
Difenylamin
Formel: (C₆H₅)₂NH
Kokepunkt: 302 °C (1013 hPa)
Smeltepunkt: 53…54 °C
Tetthet: 1,165 g/cm³ (20 °C)
Flash punkt: 153 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003014
CAS-nummer: 122-39-4
EINECS: 204-539-4
FN: 3077
ADR: 9,III
Merck Indeks: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR