1-Amino-2-methyl-9,10-anthraquinone

Leverandør: EHRENSTORFER

C12972111
EHERC12972111EA 3175 NOK
EHERC12972111
1-Amino-2-methyl-9,10-anthraquinone
1-Amino-2-methyl-9,10-anthraquinone
Formel: C₁₅H₁₁NO₂
MDL nummer: MFCD00001220
CAS-nummer: 82-28-0
EINECS: 201-408-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR