2-Ethylhexyl diphenyl phosphate

Leverandør: EHRENSTORFER

C13342300
EHERC13342300EA 3225 NOK
EHERC13342300
2-Ethylhexyl diphenyl phosphate
2-Ethylhexyl diphenyl phosphate
Formel: C₂₀H₂₇O₄P
MDL nummer: MFCD00059949
CAS-nummer: 1241-94-7
EINECS: 214-987-2
FN: 3082
ADR: 9,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR