Folic acid

Leverandør: EHRENSTORFER

Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

C13888000
EHERC13888000EA 1550 NOK
EHERC13888000
Folic acid
Folic acid
Formel: C₁₉H₁₉N₇O₆
MDL nummer: MFCD00079305
CAS-nummer: 59-30-3
EINECS: 200-419-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR