Genistein

Leverandør: EHRENSTORFER

Synonymer: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

C13999800
EHERC13999800EA 6450 NOK
EHERC13999800
Genistein
Genistein
Formel: C₁₅H₁₀O₅
Smeltepunkt: 297…298 °C
MDL nummer: MFCD00016952
CAS-nummer: 446-72-0
EINECS: 207-174-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR