Butyl 4-hydroksybenzoat

Leverandør: EHRENSTORFER

Synonymer: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

C14228780
EHERC14228780EA 2225 NOK
EHERC14228780
Butyl 4-hydroksybenzoat
Butyl 4-hydroksybenzoat
Formel: C₁₁H₁₄O₃
MW: 194,23 g/mol
Kokepunkt: 156…157 °C (3,5 mmHg)
Smeltepunkt: 73 °C
Tetthet: 1,23 g/cm³ (20 °C)
MDL nummer: MFCD00016478
CAS-nummer: 94-26-8
EINECS: 202-318-7
Merck Indeks: 13,01583

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR