Maleinsyre

Leverandør: EHRENSTORFER
Warning

C14726000
EHERC14726000EA 1600 NOK
EHERC14726000
Maleinsyre
Maleinsyre
Formel: C₄H₄O₄
MW: 116,07 g/mol
Kokepunkt: 256 °C (1013 hPa)
Smeltepunkt: 130…131 °C
Tetthet: 1,59 g/cm³ (20 °C)
Flash punkt: 127 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00063177
CAS-nummer: 110-16-7
EINECS: 203-742-5
FN: 3261
ADR: 8,III
Merck Indeks: 13,05726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR