6-Methoxy-m-toluidine

Leverandør: EHRENSTORFER
Danger

Synonymer: 3-Amino-4-methoxytoluene, 2-Amino-4-methylanisole, 1-Amino-2-methoxy-5-methylbenzene, 5-Methyl-o-anisidine, p-Cresidine, 2-Methoxy-5-methylaniline

C15081000
EHERC15081000EA 1700 NOK
EHERC15081000
6-Methoxy-m-toluidine
6-Methoxy-m-toluidine
Formel: C₈H₁₁NO
Kokepunkt: 235 °C (1013 hPa)
Smeltepunkt: 55,5 °C
Tetthet: 1 g/cm³ (20 °C)
MDL nummer: MFCD00007815
CAS-nummer: 120-71-8
EINECS: 204-419-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR