3-Nitrotoluen

Leverandør: EHRENSTORFER

Synonymer: m-Nitrotoluene

C15615300
EHERC15615300EA 1075 NOK
EHERC15615300
3-Nitrotoluen
3-Nitrotoluen
Formel: C₇H₇NO₂
MW: 137,14 g/mol
Kokepunkt: 230 °C (1013 hPa)
Smeltepunkt: 15,5 °C
Tetthet: 1,1571 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007265
CAS-nummer: 99-08-1
EINECS: 202-728-6
FN: 1664
ADR: 6.1,II
Merck Indeks: 12,06748

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR