6-Propyl-2-thiouracil

Leverandør: EHRENSTORFER
Warning

Synonymer: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

C16530500
EHERC16530500EA 1600 NOK
EHERC16530500
6-Propyl-2-thiouracil
6-Propyl-2-thiouracil
Formel: C₇H₁₀N₂OS
MW: 170,24 g/mol
Kokepunkt: 448 °C (1013 hPa)
Smeltepunkt: 215…218 °C
MDL nummer: MFCD00006041
CAS-nummer: 51-52-5
EINECS: 200-103-2
Merck Indeks: 12,08054

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR