Rifampicin

Leverandør: EHRENSTORFER

Synonymer: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP

C16814500
EHERC16814500EA 1625 NOK
EHERC16814500
Rifampicin
Rifampicin
Formel: C₄₃H₅₈N₄O₁₂
MW: 822,95 g/mol
Smeltepunkt: 183…188 °C
MDL nummer: MFCD00151389
CAS-nummer: 13292-46-1
EINECS: 236-312-0
Merck Indeks: 12,08382

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR