Dimethyl succinate

Leverandør: EHRENSTORFER
Warning

Synonymer: DBE-4 dibasic ester, Succinic acid dimethyl ester, 1,4-Butanedioic acid dimethyl ester

C16985500
EHERC16985500EA 1550 NOK
EHERC16985500
Dimethyl succinate
Dimethyl succinate
Formel: C₆H₁₀O₄
Kokepunkt: 200 °C (1013 hPa)
Smeltepunkt: 18…19 °C
Tetthet: 1,117 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 12,09038

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR