Tributyltin klorid

Leverandør: EHRENSTORFER
Danger

Synonymer: Chlorotributylstannane, Tri-n-butyltin chloride, TBTC

C17150000
EHERC17150000EA 2150 NOK
EHERC17150000
Tributyltin klorid
Tributyltin klorid
Formel: C₁₂H₂₇ClSn
MW: 325,51 g/mol
Kokepunkt: 171…173 °C (25 mmHg)
Smeltepunkt: –9 °C
Tetthet: 1,2 g/cm³ (20 °C)
Flash punkt: >100 °C
MDL nummer: MFCD00000521
CAS-nummer: 1461-22-9
EINECS: 215-958-7
FN: 2788
ADR: 6.1,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR