p-Toluidin

Leverandør: EHRENSTORFER
Danger

Synonymer: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine

C17595000
EHERC17595000EA 1075 NOK
EHERC17595000
p-Toluidin
p-Toluidin
Formel: C₇H₉N
Kokepunkt: 201 °C (1013 hPa)
Smeltepunkt: 43 °C
Tetthet: 1,046 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007906
CAS-nummer: 106-49-0
EINECS: 203-403-1
FN: 3451
ADR: 6.1,II
Merck Indeks: 12,09674

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR