D-(+)-Trehalose dihydrate

Leverandør: EHRENSTORFER

Synonymer: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

C17607500
EHERC17607500EA 1225 NOK
EHERC17607500
D-(+)-Trehalose dihydrate
D-(+)-Trehalose dihydrate
Formel: C₁₂H₂₂O₁₁·2H₂O
MW: 378,33 g/mol
Smeltepunkt: 97…99 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00071594
CAS-nummer: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR