Tributylamin

Leverandør: EHRENSTORFER
Danger

C17667500
EHERC17667500EA 1125 NOK
EHERC17667500
Tributylamin
Tributylamin
Formel: C₁₂H₂₇N
Kokepunkt: 216 °C (1013 hPa)
Smeltepunkt: –70 °C
Tetthet: 0,7771 g/cm³ (20 °C)
Flash punkt: 63 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009431
CAS-nummer: 102-82-9
EINECS: 203-058-7
FN: 2542
ADR: 6.1,II
Merck Indeks: 13,09691

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR