2,3,6-Trichloroanisole

Leverandør: EHRENSTORFER

C17713600
EHERC17713600EA 1875 NOK
EHERC17713600
2,3,6-Trichloroanisole
2,3,6-Trichloroanisole
Formel: C₇H₅Cl₃O
MW: 211,47 g/mol
Kokepunkt: 227 °C (1013 hPa)
Smeltepunkt: 45 °C
Tetthet: 1,42 g/cm³ (20 °C)
CAS-nummer: 50375-10-5
EINECS: 256-561-9
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR