2-Tridekanon

Leverandør: EHRENSTORFER

C17818500
EHERC17818500EA 1400 NOK
EHERC17818500
2-Tridekanon
2-Tridekanon
Formel: C₁₃H₂₆O
MW: 198,35 g/mol
Kokepunkt: 263 °C (1013 hPa)
Smeltepunkt: 25…27 °C
Tetthet: 0,82 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008968
CAS-nummer: 593-08-8
EINECS: 209-784-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR