1,2,4-Trimetoksybensen

Leverandør: EHRENSTORFER

C17876420
EHERC17876420EA 2025 NOK
EHERC17876420
1,2,4-Trimetoksybensen
1,2,4-Trimetoksybensen
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Kokepunkt: 234 °C (1013 hPa)
Smeltepunkt: 19…21 °C
Tetthet: 1,126 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008360
CAS-nummer: 135-77-3
EINECS: 205-219-7
Merck Indeks: 13,01073

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR