Tris(2-ethylhexyl) phosphate

Leverandør: EHRENSTORFER

C17894400
EHERC17894400EA 1375 NOK
EHERC17894400
Tris(2-ethylhexyl) phosphate
Tris(2-ethylhexyl)phosphate
Formel: C₂₄H₅₁O₄P
MDL nummer: MFCD00009491
CAS-nummer: 78-42-2
EINECS: 201-116-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR