Ethyl undecanoate

Leverandør: EHRENSTORFER

Synonymer: Undecylic acid ethyl ester, Undecanoic acid ethyl ester

C17896420
EHERC17896420EA 1375 NOK
EHERC17896420
Ethyl undecanoate
Ethyl undecanoate
Formel: C₁₃H₂₆O₂
MW: 214,35 g/mol
Kokepunkt: 258 °C (1013 hPa)
Smeltepunkt: –14,7 °C
Tetthet: 0,86 g/cm³ (20 °C)
MDL nummer: MFCD00008958
CAS-nummer: 627-90-7
EINECS: 211-018-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR