p-Xylen

Leverandør: EHRENSTORFER
Danger

Synonymer: 1,4-Dimethylbenzene

C17945200
EHERC17945200EA 1975 NOK
EHERC17945200
p-Xylen
p-Xylen
Formel: C₆H₄(CH₃)₂
Kokepunkt: 138,3 °C (1013 hPa)
Smeltepunkt: 13 °C
Tetthet: 1,096 g/cm³ (20 °C)
Flash punkt: 25 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008556
CAS-nummer: 106-42-3
EINECS: 203-396-5
FN: 1307
ADR: 3,III
Merck Indeks: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR