Phenazine

Leverandør: EHRENSTORFER

C20921500
EHERC20921500EA 1725 NOK
EHERC20921500
Phenazine
Phenazine
Formel: C₁₂H₈N₂
MW: 180,21 g/mol
Kokepunkt: 339 °C (1013 hPa)
Smeltepunkt: 172…174 °C
Tetthet: 1,26 g/cm³ (22 °C)
MDL nummer: MFCD00005023
CAS-nummer: 92-82-0
EINECS: 202-193-9
Merck Indeks: 12,07359

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR