cis-3-Hexenyl acetate

Leverandør: EHRENSTORFER

CA14202500
EHERCA14202500EA 2775 NOK
EHERCA14202500
cis-3-Hexenyl acetate
cis-3-Hexenyl acetate
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
MDL nummer: MFCD00036563
CAS-nummer: 3681-71-8
EINECS: 222-960-1
FN: 3272
ADR: 3,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR