Organic reference standard, Dihexyl phthalate 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Leverandør: EHRENSTORFER
L16173200CY
EHERL16173200CYEA 1700 NOK
EHERL16173200CY
Organic reference standard, Dihexyl phthalate 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Standarder Organiske standarder
Bestill nå
SPESIFIKASJONER
Beskrivelse Dihexyl phthalate
CAS-nr 84-75-3
Formel C₂₀H₃₀O₄