Pesticide reference standard, 2,6-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer

Leverandør: EHRENSTORFER

XA12453000ME
EHERXA12453000MEEA 530 NOK
EHERXA12453000ME
Pesticide reference standard, 2,6-Dichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Standarder Organiske standarder

Bestill nå

SPESIFIKASJONER

Beskrivelse 2,6-Dichlorophenol
CAS-nr 87-65-0
Formel C₆H₄Cl₂O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR