Chemical Standards Kit for Triple Quad Instruments
Leverandør: EKSIGENT
4406127
EKSI4406127EA 0 NOK
EKSI4406127
Chemical Standards Kit for Triple Quad Instruments
Standarder Fargereferanse og spekroskopistandarer
Standards chemical kit with low and high concentrations of polypropylene glycol polymers (PPGs).
  • Ensure data consistency - avoid risking the integrity of your workflow
  • Save time and resources
  • Maintain peak performance - troubleshoot your mass spectrometer in minutes
  • Qualify instrument for IQ/OQ/PQ
This standards kit ensures mass spectrometer to turned for optimal performance and delivers reproducible results.
Leveringsinformasjon: This kit includes all solutions necessary for installation and calibration.
NB: For research use only. Not for use in diagnostic procedures.