Test sieves, 200×25 mm
Leverandør: ENDECOTTS
200SAW.600H
ENDE200SAW.600HEA 2775 NOK
ENDE200SAW.600H
Test sieves, 200×25 mm
Sikter Sikter, test
These test sieves have a brass frame and stainless steel mesh.
Sertifiseringer: According to ASTM E 11-95
Leveringsinformasjon: Each sieve is delivered with an individual quality certificate.