Orbipac CPS51 pH-ORP Sensor
Leverandør: Endress+Hauser
CPS51-1EK2FDA
ENDRCPS51-1EK2FDAEA 3475 NOK
ENDRCPS51-1EK2FDA
Orbipac CPS51 pH-ORP Sensor
Elektroder pH-elektroder
pH electrode for handheld and laboratory measuring devices.
  • Fixed cable: 1,5 m
  • CPS51 excluding temperature sensor: 1EK2FDA
  • Working limit: Pressureless
Sertifiseringer: EN IEC 63000:2018.