Nitrogen spray gun
Leverandør: ENTEGRIS
421-42-11 421-42-21 421-42-31
ENTE421-42-11EA 0 NOK
ENTE421-42-11 ENTE421-42-21 ENTE421-42-31
Nitrogen spray gun
Sprøytepistol
Long lasting and reliable PFA nitrogen spray guns with 0,5 µm PTFE membrane for use in high purity and corrosive chemical applications.
  • Integral filter enables air to be filtered before nitrogen is dispensed
  • Vented cap/nozzle meets OSHA safety specifications
  • PFA construction enables use in high purity and corrosive applications
  • Colored caps enable visual chemical differentiation at a glance
Nitrogen cap/nozzle prevents pressure build-up if outlet is blocked.
Orifice 4,8 mm
Max. pressure 75 psig (517 kPa)
Connection ¹/₄" FNPT
Tilbehørs informasjon:More Product Information