Miscellaneous device carriers A118
Leverandør: ENTEGRIS
A118-01-0215
ENTEA118-01-0215EA 0 NOK
ENTEA118-01-0215
Miscellaneous device carriers A118
Wafer håndtering Wafer bærere
Carriers for handling miscellaneous devices.
  • Pocket size (L×W×H): 25,4×7,9×9,5 mm (1,00×0,31×0,38")
  • Overall size (L×W×H): 76×74×15 mm (3,0×2,9×0,6")
  • Wash openings in each pocket
  • Capacity: 18