Wafer carrier handles for 76,2 mm (3") wafer handling
Leverandør: ENTEGRIS
A82-03-0215
ENTEA82-03-0215EA 0 NOK
ENTEA82-03-0215
Wafer carrier handles for 76,2 mm (3") wafer handling
Wafer håndtering Wafer bærere
  • Attaches to the notches on both end walls of carrier
  • Overall size: 165×91×157 mm