Sample dipper with v bottom
Leverandør: ENTEGRIS
D11-0215 D14-0215 D15-0215
ENTED11-0215EA 0 NOK
ENTED11-0215 ENTED14-0215 ENTED15-0215
Sample dipper with v bottom
Prøvetakere Prøveøser
Holds a wafer or a square plate.
  • Slot has 'V' bottom
  • Clip on handle slides down to hold substrates in place
  • Hold one wafer or one square plate