Round process tank
Leverandør: ENTEGRIS
E14-35-01-0215
ENTEE14-35-01-0215EA 0 NOK
ENTEE14-35-01-0215
Round process tank
Store beholdere Store beholdere, sylindriske
Injection molded round tank, in PFA material.
  • Tanks should be properly supported in heated applications
  • Maximum recommended temperature limit for PFA tanks is 180 °C