GateKeeper® GPU 17M 12R Gas Purifier
Leverandør: ENTEGRIS
GPUS17MFIX12R0
ENTEGPUS17MFIX12R0EA 0 NOK
ENTEGPUS17MFIX12R0
GateKeeper® GPU 17M 12R Gas Purifier
Filter Gassfilter
Entegris’ GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime, and final purity in microelectronics applications where high-purity gas is critical. Leveraging our expertise in material science, we have developed a new generation of IX gas purification products that deliver unsurpassed value and performance.
  • Purity control below part-per-billion (ppb) levels
  • Low pressure drop through the purifier
  • Cost effective
  • Regenerable - eliminates disposal issues and costs
  • Long lifetime - improves cost of ownership
  • Safe and simple to use