GateKeeper® GPU 5000 Gas Purifiers
Leverandør: ENTEGRIS
GPUS5000FEXR02
ENTEGPUS5000FEXR02EA 0 NOK
ENTEGPUS5000FEXR02
GateKeeper® GPU 5000 Gas Purifiers
Filter Gassfilter
GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime and final purity. Leveraging its expertise in material science, Entegris has developed a new generation of purification products that deliver unsurpassed value and performance.
  • Purity control to ppt levels
  • Low pressure drop across the purifier
  • Regenerable
  • Long lifetime
  • Use ambient temperature purification – no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)
The model 5000 is designed for peak flow rates up to 500 slpm.