Inserts for trays H 44 series
Leverandør: ENTEGRIS
H44-001B H44-001D
ENTEH44-001BEA 0 NOK
ENTEH44-001B ENTEH44-001D
Inserts for trays H 44 series
Wafer håndtering Wafer, brett
Nest between H 44 trays and/or covers.
  • Precut to tray size
  • Inserts provide a softer surface providing protection for more sensitive components
  • Inserts are not intended to limit part migration
  • Inserts with antistatic coatings are not recommended for use when storing devices for extended periods