Wafer shipping system, horizontal, SmartStack®
Leverandør: ENTEGRIS
HWS-KIT-CUSHE
ENTEHWS-KIT-CUSHEEA 0 NOK
ENTEHWS-KIT-CUSHE
Wafer shipping system, horizontal, SmartStack®
Wafer håndtering Wafer forsendere
SmartStack® horizontal wafer shipping system is designed to reduce wafer breakage, die bond corrosion and particle contamination. These shipping system is designed to utilize common secondary packaging across wafer sizes, and the result is reduction in the number of pacvkaging configurations and wafer packing process. With additional layer of protection it is easy to maintain wafer integrity during transport and shipping.
  • Multiple configurations are common across wafer sizes as 1-pack system holds one canister and 2-pack hold two canisters
  • Passes ISTA-2A