Single wafer shipping boxes for 200 mm wafer handling, Solopak® series
Leverandør: ENTEGRIS
Z200-0203_U
ENTEZ200-0203_UEA 0 NOK
ENTEZ200-0203_U
Single wafer shipping boxes for 200 mm wafer handling, Solopak® series
Wafer håndtering Wafer forsendere
Polycarbonate, clear.
  • Material allows visual inspection without opening
  • Prevent wafer rotation and particle generation
  • Large top surface provide space for labelling
  • Pin-in-socket allows the shipper to be opened horizontally
It holds one wafer facedown or faceup in a conical pocket, contacting wafer edges alone.
Dimensions: 232×218×19 mm