RIPA cell lysis buffer 2
Leverandør: ENZO LIFE SCIENCES
ADI801284
ENZOADI801284EA 3675 NOK
ENZOADI801284
RIPA cell lysis buffer 2
Buffere Buffere, ekstrasjons
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Formulering Liquid, 50 mm TRIS-hydrogen chloride, pH 7,4, containing 150 mm sodium chloride, 1 mm EDTA, 1 mm EGTA, 1% Triton X-100, 1% sodium deoxycholate and 0,1% SDS