Cell lysis buffer, RIPA
Leverandør: ENZO LIFE SCIENCES
ADI801496
ENZOADI801496EA 2950 NOK
ENZOADI801496
Cell lysis buffer, RIPA
Buffere Buffere, ekstrasjons
The RIPA liquid cell lysis buffer composes of 50 mM TRIS-hydrogen chloride, pH 7,4, containing 150 mM sodium chloride, 1 mM EDTA, 1 mM EGTA, 1,2% Triton X-100, 0,5% sodium deoxycholate, and 0,1% SDS.
Storage temperature: 4 °C
NB: RUO - Research use only.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Formulering Liquid. 50 mM Tris, containing 150 mM NaCl, 1 mM EDTA, 1 mM EGTA, 1,2% Tergitol 15-S-9, 0,5% sodium deoxycholate, and 0,1% SDS
Fraktforhold 20
Lagringstemp 4 °C (Long term storage)