Stop solution 2
Leverandør: ENZO LIFE SCIENCES
ADI801804
ENZOADI801804EA 1100 NOK
ENZOADI801804
Stop solution 2
Assay ELISAs
Acid stop solution (1 N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions.
Each bottle is sufficient for use with 5×96 wells microtiter plates.
Bestillingsinformasjon: Additional assay development reagents are available as companion products to IMMUNOSET® products.